missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012813
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012813 1 mL
111012814 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 111012813 missing translation for 'mfr' Merck Emd Millipore missing translation for 'supplierNo' 567531MLF

2-Phenoxyethanol is a compound, which belongs to the class of volatile organicshaving a faint rose like aromatic odor. It may find applications as a solvent in ball-point inks, resins and in organic synthesis. It may also be used in cosmetics as an antimicrobial agent and preservative.

Featured Industries: Environmental

Specifications

CAS 122-99-6
Density 1.107 g/mL (at 20°C)
Flash Point 121°C
Physical Form Neat
Linear Formula C6H5 OCH2 CH2 OH
MDL Number MFCD00002857
Melting Point 11°C to 13°C
Boiling Point 244°C to 246°C
Synonym Ethylene glycol monophenyl ether; PHE; PHE-G; PHE-S; PhG; Phenylglycol
Type Analytical Standard
Quantity 1 mL
Formula Weight 138.16
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less