Learn More
2-Phenoxybenzoic acid, 98%
CAS: 2243-42-7 | C13H10O3 | 214.22 g/mol
Supplier: Thermo Scientific Chemicals A1051514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Phenoxybenzoic acid | |
| 111°C to 115°C | |
| MFCD00002429 | |
| 978452 | |
| PKRSYEPBQPFNRB-UHFFFAOYSA-N | |
| 2-phenoxybenzoic acid | |
| 75237 | |
| 214.22 |
| 2243-42-7 | |
| C13H10O3 | |
| 25 g | |
| o-phenoxybenzoic acid, benzoic acid, 2-phenoxy, benzoic acid, o-phenoxy, phenoxybenzoic acid, ortho-phenoxybenzoic acid, 2-phenoxybenzoicacid, 2-carboxydiphenyl ether, 2-phenoxy-benzoic acid, o-phenoxybenzoic acid, o-carboxydiphenyl ether | |
| C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)O | |
| 214.22 | |
| CHEBI:72636 | |
| 98% |
Chemical Identifiers
| 2243-42-7 | |
| 214.22 | |
| PKRSYEPBQPFNRB-UHFFFAOYSA-N | |
| 75237 | |
| 2-phenoxybenzoic acid |
| C13H10O3 | |
| MFCD00002429 | |
| o-phenoxybenzoic acid, benzoic acid, 2-phenoxy, benzoic acid, o-phenoxy, phenoxybenzoic acid, ortho-phenoxybenzoic acid, 2-phenoxybenzoicacid, 2-carboxydiphenyl ether, 2-phenoxy-benzoic acid, o-phenoxybenzoic acid, o-carboxydiphenyl ether | |
| CHEBI:72636 | |
| C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-811-5
RTECSNumber : DH6293560
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only