missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzoic acid, 98%

Catalog No. p-7080809
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1051503 1 g
AAA1051514 25 g
AAA1051522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1051503 Supplier Thermo Scientific Chemicals Supplier No. A1051503
Only null left

CAS: 2243-42-7 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2243-42-7
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002429
InChI Key PKRSYEPBQPFNRB-UHFFFAOYSA-N
Synonym o-phenoxybenzoic acid, benzoic acid, 2-phenoxy, benzoic acid, o-phenoxy, phenoxybenzoic acid, ortho-phenoxybenzoic acid, 2-phenoxybenzoicacid, 2-carboxydiphenyl ether, 2-phenoxy-benzoic acid, o-phenoxybenzoic acid, o-carboxydiphenyl ether
PubChem CID 75237
ChEBI CHEBI:72636
IUPAC Name 2-phenoxybenzoic acid
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)O

Specifications

Melting Point 111°C to 115°C
Quantity 1 g
Beilstein 978452
Formula Weight 214.22
Percent Purity 98%
Chemical Name or Material 2-Phenoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-811-5
RTECSNumber DH6293560
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only