missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzeneboronic acid, 98%

Catalog No. p-7035546
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5326403 1 g
AAH5326406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5326403 Supplier Thermo Scientific Chemicals Supplier No. H5326403

CAS: 108238-09-1 | C12H11BO3 | 214.03 g/mol

Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 108238-09-1
Molecular Formula C12H11BO3
Molecular Weight (g/mol) 214.03
MDL Number MFCD01001592
InChI Key AVOWPOFIQZSVGV-UHFFFAOYSA-N
Synonym 2-phenoxybenzeneboronic acid, 2-phenoxyphenyl boronic acid, 2-phenoxyphenyl boranediol, 2-phenoxy phenylboronic acid, boronic acid, 2-phenoxyphenyl, pubchem9521, acmc-1bnrk, ksc174e3r, 2-phenoxy-phenyl boronic acid, 2-phenoxyphenylboronicacid
PubChem CID 2773559
IUPAC Name (2-phenoxyphenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1OC1=CC=CC=C1

Specifications

Melting Point 116°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 214.03
Percent Purity 98%
Chemical Name or Material 2-Phenoxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only