missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyaniline, 98%

CAS: 2688-84-8 | C12H11NO | 185.226 g/mol

$84.90 - $211.84

Chemical Identifiers

CAS 2688-84-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00035765
InChI Key NMFFUUFPJJOWHK-UHFFFAOYSA-N
Synonym 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
PubChem CID 75899
IUPAC Name 2-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0555814
View Documents
Thermo Scientific Chemicals
L0555814
25 g
Each for $84.90
Only null left
 
AAL0555822
View Documents
Thermo Scientific Chemicals
L0555822
100 g
Each for $211.84
Only null left
 
Description

Description

2-Phenoxyaniline serves as an antiinflammatory agent that inhibits preferentially COX-2 over COX-1. It is used in the preparation of sodium primary amide complex, 4-methyl-2-(2-phenoxyphenyl)azo-phenol and 2-acetoaminodiphenyl ether. Further, it is used to form complexes with beta-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxyaniline serves as an antiinflammatory agent that inhibits preferentially COX-2 over COX-1. It is used in the preparation of sodium primary amide complex, 4-methyl-2-(2-phenoxyphenyl)azo-phenol and 2-acetoaminodiphenyl ether. Further, it is used to form complexes with beta-cyclodextrin.

Solubility
Slightly soluble in chloroform and methanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

2688-84-8
185.226
NMFFUUFPJJOWHK-UHFFFAOYSA-N
75899
C1=CC=C(C=C1)OC2=CC=CC=C2N
C12H11NO
MFCD00035765
2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
2-phenoxyaniline

Specifications

2688-84-8
170°C (18 mmHg)
C12H11NO
25 g
2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
NMFFUUFPJJOWHK-UHFFFAOYSA-N
2-phenoxyaniline
75899
99%
46°C to 48°C
>110°C (230°F)
MFCD00035765
777766
Slightly soluble in chloroform and methanol.
C1=CC=C(C=C1)OC2=CC=CC=C2N
185.226
185.23
2-Phenoxyaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 220-254-8

RTECSNumber : BY7940000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only