missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyaniline, 98%

Catalog No. AAL0555814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0555814 25 g
AAL0555822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0555814 Supplier Thermo Scientific Chemicals Supplier No. L0555814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2688-84-8 | C12H11NO | 185.226 g/mol

2-Phenoxyaniline serves as an antiinflammatory agent that inhibits preferentially COX-2 over COX-1. It is used in the preparation of sodium primary amide complex, 4-methyl-2-(2-phenoxyphenyl)azo-phenol and 2-acetoaminodiphenyl ether. Further, it is used to form complexes with beta-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxyaniline serves as an antiinflammatory agent that inhibits preferentially COX-2 over COX-1. It is used in the preparation of sodium primary amide complex, 4-methyl-2-(2-phenoxyphenyl)azo-phenol and 2-acetoaminodiphenyl ether. Further, it is used to form complexes with beta-cyclodextrin.

Solubility
Slightly soluble in chloroform and methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2688-84-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00035765
InChI Key NMFFUUFPJJOWHK-UHFFFAOYSA-N
Synonym 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
PubChem CID 75899
IUPAC Name 2-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2N

Specifications

Melting Point 46°C to 48°C
Boiling Point 170°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 777766
Solubility Information Slightly soluble in chloroform and methanol.
Formula Weight 185.23
Percent Purity 99%
Chemical Name or Material 2-Phenoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-254-8
RTECSNumber BY7940000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only