missing translation for 'onlineSavingsMsg'
Learn More

2-p-Terphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T24121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24121G Supplier TCI America Supplier No. T24121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 663954-31-2
Molecular Formula C18H15BO2
Molecular Weight (g/mol) 274.13
MDL Number MFCD08669638
InChI Key SNYOIUKSBGFPSV-UHFFFAOYSA-N
Synonym 2-p-terphenylboronic acid, 1,1':4',1-terphenyl-2-ylboronic acid, 1,1':4',1-terphenyl-2-boronic acid, 2-4-biphenyl phenylboronic acid, boronic acid, 1,1':4',1-terphenyl-2-yl, 2-1,1'-biphenyl-4-yl phenylboronic acid, 2-p-terphenylboronicacid, 2-4-phenylphenyl phenylboronic acid, boronic acid,b-1,1':4',1-terphenyl-2-yl, 1∼1∼,2∼1∼:2∼4∼,3∼1∼-terphenyl-1∼2∼-ylboronic acid
PubChem CID 22168980
IUPAC Name (2-{[1,1'-biphenyl]-4-yl}phenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 274.13
Physical Form Crystalline Powder
Chemical Name or Material 2-p-Terphenylboronic Acid (contains varying amounts of Anhydride)