accessibility menu, dialog, popup

UserName

CAS RN 663954-31-2

IUPAC Name: (2-{[1,1'-biphenyl]-4-yl}phenyl)boronic acid
Synonyms: 2-{[1,1'-biphenyl]-4-yl}phenylboronic acid
Molecular Weight (g/mol): 274.13
Molecular Formula: C18H15BO2
InChi Key: SNYOIUKSBGFPSV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1C1=CC=C(C=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results