missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-5-azaspiro[3.4]octane oxalate, 96%

CAS: 1389264-18-9 | C8H13NO5 | 203.194 g/mol

Supplier:  Thermo Scientific Chemicals H57854ME

 View more versions of this product

Catalog No. AAH57854ME

Only null left
Explore available promotions

Description

Description

Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

Solubility
Decomposes in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
Specifications

Specifications

2-Oxa-5-azaspiro[3.4]octane oxalate
C8H13NO5
500 mg
Decomposes in water.
C1CC2(COC2)NC1.C(=O)(C(=O)O)O
203.194
203.19
1389264-18-9
MFCD18325162
2-oxa-5-azaspiro 3.4 octane oxalate, 2-oxa-5-aza-spiro 3.4 octane oxalate, 2-oxa-5-azaspiro 3.4 octane oxalate salt, 2-oxa-5-azaspiro 3.4 octane, hemioxalate salt, 2-oxa-5-azaspiro 3.4 octane; oxalic acid
JFOZNINEJYPQQK-UHFFFAOYSA-N
2-oxa-5-azaspiro[3.4]octane;oxalic acid
72207353
96%

Chemical Identifiers

1389264-18-9
203.194
JFOZNINEJYPQQK-UHFFFAOYSA-N
72207353
C1CC2(COC2)NC1.C(=O)(C(=O)O)O
C8H13NO5
MFCD18325162
2-oxa-5-azaspiro 3.4 octane oxalate, 2-oxa-5-aza-spiro 3.4 octane oxalate, 2-oxa-5-azaspiro 3.4 octane oxalate salt, 2-oxa-5-azaspiro 3.4 octane, hemioxalate salt, 2-oxa-5-azaspiro 3.4 octane; oxalic acid
2-oxa-5-azaspiro[3.4]octane;oxalic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only