missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-5-azaspiro[3.4]octane oxalate, 96%

Catalog No. p-7038805
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57854MD 250 mg
AAH57854ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57854MD Supplier Thermo Scientific Chemicals Supplier No. H57854MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1389264-18-9 | C8H13NO5 | 203.194 g/mol

Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

Solubility
Decomposes in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive

Chemical Identifiers

CAS 1389264-18-9
Molecular Formula C8H13NO5
Molecular Weight (g/mol) 203.194
MDL Number MFCD18325162
InChI Key JFOZNINEJYPQQK-UHFFFAOYSA-N
Synonym 2-oxa-5-azaspiro 3.4 octane oxalate, 2-oxa-5-aza-spiro 3.4 octane oxalate, 2-oxa-5-azaspiro 3.4 octane oxalate salt, 2-oxa-5-azaspiro 3.4 octane, hemioxalate salt, 2-oxa-5-azaspiro 3.4 octane; oxalic acid
PubChem CID 72207353
IUPAC Name 2-oxa-5-azaspiro[3.4]octane;oxalic acid
SMILES C1CC2(COC2)NC1.C(=O)(C(=O)O)O

Specifications

Quantity 250 mg
Solubility Information Decomposes in water.
Formula Weight 203.19
Percent Purity 96%
Chemical Name or Material 2-Oxa-5-azaspiro[3.4]octane oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only