missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-O-Methyluridine 98.0+%, TCI America™
$121.16 - $363.49
Chemical Identifiers
| CAS | 2140-76-3 |
|---|---|
| Molecular Formula | C10H14N2O6 |
| Molecular Weight (g/mol) | 258.23 |
| MDL Number | MFCD00056054 |
| InChI Key | SXUXMRMBWZCMEN-ZOQUXTDFSA-N |
| Synonym | 2'-o-methyluridine, uridine, 2'-o-methyl, 1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, o 2'-methyluridine, 2-o-methyluridine, 2'-o-methyl uridine, unii-399vzb6tmb, 399vzb6tmb, 2'-o-methyl-uridine, um |
| PubChem CID | 102212 |
| ChEBI | CHEBI:19227 |
| IUPAC Name | 1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidine-2,4-dione |
| SMILES | COC1C(C(OC1N2C=CC(=O)NC2=O)CO)O |
Chemical Identifiers
| 2140-76-3 | |
| 258.23 | |
| SXUXMRMBWZCMEN-ZOQUXTDFSA-N | |
| 102212 | |
| 1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidine-2,4-dione |
| C10H14N2O6 | |
| MFCD00056054 | |
| 2'-o-methyluridine, uridine, 2'-o-methyl, 1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, o 2'-methyluridine, 2-o-methyluridine, 2'-o-methyl uridine, unii-399vzb6tmb, 399vzb6tmb, 2'-o-methyl-uridine, um | |
| CHEBI:19227 | |
| COC1C(C(OC1N2C=CC(=O)NC2=O)CO)O |
Specifications
| 2140-76-3 | |
| White | |
| MFCD00056054 | |
| 2'-o-methyluridine, uridine, 2'-o-methyl, 1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, o 2'-methyluridine, 2-o-methyluridine, 2'-o-methyl uridine, unii-399vzb6tmb, 399vzb6tmb, 2'-o-methyl-uridine, um | |
| COC1C(C(OC1N2C=CC(=O)NC2=O)CO)O | |
| 258.23 | |
| CHEBI:19227 | |
| ≥98.0% (HPLC,T) | |
| 2′-O-Methyluridine |
| 161°C | |
| C10H14N2O6 | |
| 1 g | |
| SXUXMRMBWZCMEN-ZOQUXTDFSA-N | |
| 1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidine-2,4-dione | |
| 102212 | |
| 258.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator