missing translation for 'onlineSavingsMsg'
Learn More

2'-O-Methyluridine 98.0+%, TCI America™

Supplier:  TCI America M22905G

 View more versions of this product

Catalog No. M22905G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2′-O-Methyluridine
161°C
C10H14N2O6
5 g
SXUXMRMBWZCMEN-ZOQUXTDFSA-N
1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidine-2,4-dione
102212
258.23
Crystalline Powder
2140-76-3
White
MFCD00056054
2'-o-methyluridine, uridine, 2'-o-methyl, 1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, o 2'-methyluridine, 2-o-methyluridine, 2'-o-methyl uridine, unii-399vzb6tmb, 399vzb6tmb, 2'-o-methyl-uridine, um
COC1C(C(OC1N2C=CC(=O)NC2=O)CO)O
258.23
CHEBI:19227
≥98.0% (HPLC,T)

Chemical Identifiers

2140-76-3
258.23
SXUXMRMBWZCMEN-ZOQUXTDFSA-N
102212
1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidine-2,4-dione
C10H14N2O6
MFCD00056054
2'-o-methyluridine, uridine, 2'-o-methyl, 1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, o 2'-methyluridine, 2-o-methyluridine, 2'-o-methyl uridine, unii-399vzb6tmb, 399vzb6tmb, 2'-o-methyl-uridine, um
CHEBI:19227
COC1C(C(OC1N2C=CC(=O)NC2=O)CO)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions