missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Norbornanone 98.0+%, TCI America™
Chemical Identifiers
| CAS | 497-38-1 |
|---|---|
| Molecular Formula | C7H10O |
| Molecular Weight (g/mol) | 110.156 |
| MDL Number | MFCD00074823 |
| InChI Key | KPMKEVXVVHNIEY-UHFFFAOYSA-N |
| Synonym | norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one |
| PubChem CID | 10345 |
| IUPAC Name | bicyclo[2.2.1]heptan-3-one |
| SMILES | C1CC2CC1CC2=O |
Chemical Identifiers
| C7H10O | |
| MFCD00074823 | |
| norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one | |
| bicyclo[2.2.1]heptan-3-one |
Specifications
| 497-38-1 | |
| White | |
| C7H10O | |
| 1325 | |
| KPMKEVXVVHNIEY-UHFFFAOYSA-N | |
| bicyclo[2.2.1]heptan-3-one | |
| 10345 | |
| ≥98.0% (GC) | |
| 2-Norbornanone |
| 95°C | |
| 55°C | |
| MFCD00074823 | |
| norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one | |
| C1CC2CC1CC2=O | |
| 110.156 | |
| 110.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : RB7680000
TSCA : Yes