missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Norbornanone 98.0+%, TCI America™
Supplier: TCI America N051125G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C7H10O | |
| MFCD00074823 | |
| norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one | |
| bicyclo[2.2.1]heptan-3-one |
Specifications
| 2-Norbornanone | |
| 95°C | |
| 55°C | |
| 25 g | |
| 1325 | |
| KPMKEVXVVHNIEY-UHFFFAOYSA-N | |
| bicyclo[2.2.1]heptan-3-one | |
| 10345 | |
| ≥98.0% (GC) |
| 497-38-1 | |
| White | |
| C7H10O | |
| MFCD00074823 | |
| norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one | |
| C1CC2CC1CC2=O | |
| 110.156 | |
| 110.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : RB7680000
TSCA : Yes