missing translation for 'onlineSavingsMsg'
Learn More

2-Norbornanone 98.0+%, TCI America™

Catalog No. N05115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05115G 5 g
N051125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05115G Supplier TCI America Supplier No. N05115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 497-38-1
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00074823
InChI Key KPMKEVXVVHNIEY-UHFFFAOYSA-N
Synonym norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
PubChem CID 10345
IUPAC Name bicyclo[2.2.1]heptan-3-one
SMILES C1CC2CC1CC2=O

Specifications

Melting Point 95°C
Color White
Boiling Point 55°C
Quantity 5 g
UN Number 1325
Formula Weight 110.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Norbornanone
RTECSNumber RB7680000
TSCA Yes