missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrotoluene, 99+%

Catalog No. p-7021207
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1344622 100 g
AAA1344636 500 g
AAA134460E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1344622 Supplier Thermo Scientific Chemicals Supplier No. A1344622
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Used as an intermediate, reagent, as a precursor and as a nitrogen, carbon, energy supplement | CAS: 88-72-2 | C7H7NO2 | 137.138 g/mol

2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.

Solubility
Miscible with acetone, benzene, chloroform, diethyl ether, ethanol and petroleum ether. Immiscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, sulfuric acid, reducing agents, hydrogen and sodium.

Chemical Identifiers

CAS 88-72-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007157
InChI Key PLAZTCDQAHEYBI-UHFFFAOYSA-N
Synonym 2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
PubChem CID 6944
ChEBI CHEBI:33098
IUPAC Name 1-methyl-2-nitrobenzene
SMILES CC1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -4°C
Density 1.166
Boiling Point 224°C to 226°C
Flash Point 106°C (222°F)
Refractive Index 1.545
Quantity 100 g
UN Number UN1664
Beilstein 1907580
Merck Index 14,6650
Solubility Information Miscible with acetone,benzene,chloroform,diethyl ether,ethanol and petroleum ether; Immiscible with water.
Formula Weight 137.14
Percent Purity ≥99%
Chemical Name or Material 2-Nitrotoluene
Show More Show Less
Hazard Category H302-H340-H350-H361f
Hazard Statement GHS H Statement
H340-H350-H361-H302
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROTOLUENES, LIQUID
EINECSNumber 201-853-3
RTECSNumber XT3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only