missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroterephthalic acid 1-methyl ester, 97%

CAS: 35092-89-8 | C9H7NO6 | 225.16 g/mol

$42.01 - $140.34

Chemical Identifiers

CAS 35092-89-8
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.16
MDL Number MFCD00024510
InChI Key MIIADZYPHVTLPR-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-3-nitrobenzoic acid, 1-methyl 2-nitroterephthalate, methyl hydrogen 2-nitroterephthalate, 3-nitro-4-carbomethoxybenzoic acid, 2-nitroterephthalic acid 1-methyl ester, methyl 2-nitro-4-carboxybenzoate, methyl-2-nitro-4-carboxy-benzoate, 4-carbomethoxy-3-nitro-benzoic acid
PubChem CID 98592
IUPAC Name 4-(methoxycarbonyl)-3-nitrobenzoic acid
SMILES COC(=O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2712106
View Documents
Thermo Scientific Chemicals
H2712106
5 g
Each for $42.01
Only null left
 
AAH2712114
View Documents
Thermo Scientific Chemicals
H2712114
25 g
Each for $140.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35092-89-8
225.16
MIIADZYPHVTLPR-UHFFFAOYSA-N
98592
COC(=O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O
C9H7NO6
MFCD00024510
4-methoxycarbonyl-3-nitrobenzoic acid, 1-methyl 2-nitroterephthalate, methyl hydrogen 2-nitroterephthalate, 3-nitro-4-carbomethoxybenzoic acid, 2-nitroterephthalic acid 1-methyl ester, methyl 2-nitro-4-carboxybenzoate, methyl-2-nitro-4-carboxy-benzoate, 4-carbomethoxy-3-nitro-benzoic acid
4-(methoxycarbonyl)-3-nitrobenzoic acid

Specifications

35092-89-8
C9H7NO6
5 g
MIIADZYPHVTLPR-UHFFFAOYSA-N
4-(methoxycarbonyl)-3-nitrobenzoic acid
98592
97%
173°C to 178°C
MFCD00024510
4-methoxycarbonyl-3-nitrobenzoic acid, 1-methyl 2-nitroterephthalate, methyl hydrogen 2-nitroterephthalate, 3-nitro-4-carbomethoxybenzoic acid, 2-nitroterephthalic acid 1-methyl ester, methyl 2-nitro-4-carboxybenzoate, methyl-2-nitro-4-carboxy-benzoate, 4-carbomethoxy-3-nitro-benzoic acid
COC(=O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O
225.16
225.17
2-Nitroterephthalic acid 1-methyl ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 252-360-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only