accessibility menu, dialog, popup

UserName

CAS RN 35092-89-8

IUPAC Name: 4-(methoxycarbonyl)-3-nitrobenzoic acid
Synonyms: 4-(methoxycarbonyl)-3-nitrobenzoic acid
Molecular Weight (g/mol): 225.16
Molecular Formula: C9H7NO6
InChi Key: MIIADZYPHVTLPR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results