missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroterephthalic acid 1-methyl ester, 97%

Catalog No. AAH2712106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2712106 5 g
AAH2712114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2712106 Supplier Thermo Scientific Chemicals Supplier No. H2712106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35092-89-8 | C9H7NO6 | 225.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35092-89-8
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.16
MDL Number MFCD00024510
InChI Key MIIADZYPHVTLPR-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-3-nitrobenzoic acid, 1-methyl 2-nitroterephthalate, methyl hydrogen 2-nitroterephthalate, 3-nitro-4-carbomethoxybenzoic acid, 2-nitroterephthalic acid 1-methyl ester, methyl 2-nitro-4-carboxybenzoate, methyl-2-nitro-4-carboxy-benzoate, 4-carbomethoxy-3-nitro-benzoic acid
PubChem CID 98592
IUPAC Name 4-(methoxycarbonyl)-3-nitrobenzoic acid
SMILES COC(=O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O

Specifications

Melting Point 173°C to 178°C
Quantity 5 g
Formula Weight 225.17
Percent Purity 97%
Chemical Name or Material 2-Nitroterephthalic acid 1-methyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-360-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only