missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroso-1-naphthol 98.0+%, TCI America™

Catalog No. N02675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02675G 5 g
N026725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02675G Supplier TCI America Supplier No. N02675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132-53-6
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00003932
InChI Key SYUYTOYKQOAVDW-UHFFFAOYSA-N
Synonym 2-nitroso-1-naphthol, 2-nitrosonaphthol, 1-naphthalenol, 2-nitroso, 1-naphthol, 2-nitroso, 1,2-naphthoquinone 2-oxime, nitrosonaphthol, .beta.-nitrosonaphthol, unii-cc0s5ho41v, beta-nitroso-alpha-naphthol, 1,2-naphthalenedione, 2-oxime
PubChem CID 8592
IUPAC Name 2-nitrosonaphthalen-1-ol
SMILES OC1=C2C=CC=CC2=CC=C1N=O

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 173.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitroso-1-naphthol
RTECSNumber QL4550000
TSCA Yes