missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl Phenyl Sulfide 98.0+%, TCI America™

$214.37 - $564.81

Chemical Identifiers

CAS 4171-83-9
Molecular Formula C12H9NO2S
Molecular Weight (g/mol) 231.269
InChI Key ZPWNCSAEXUDWTN-UHFFFAOYSA-N
Synonym 2-Nitrodiphenyl Sulfide, 1-Nitro-2-(phenylthio)benzene
PubChem CID 348620
IUPAC Name 1-nitro-2-phenylsulfanylbenzene
SMILES C1=CC=C(C=C1)SC2=CC=CC=C2[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N07445G
View Documents
TCI America
N07445G
5 g
Each for $214.37
Only null left
 
N074425G
View Documents
TCI America
N074425G
25 g
Each for $564.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4171-83-9
231.269
2-Nitrodiphenyl Sulfide, 1-Nitro-2-(phenylthio)benzene
1-nitro-2-phenylsulfanylbenzene
C12H9NO2S
ZPWNCSAEXUDWTN-UHFFFAOYSA-N
348620
C1=CC=C(C=C1)SC2=CC=CC=C2[N+](=O)[O-]

Specifications

4171-83-9
Yellow
5 g
ZPWNCSAEXUDWTN-UHFFFAOYSA-N
1-nitro-2-phenylsulfanylbenzene
348620
≥98.0% (GC)
2-Nitrophenyl Phenyl Sulfide
82°C
C12H9NO2S
2-Nitrodiphenyl Sulfide, 1-Nitro-2-(phenylthio)benzene
C1=CC=C(C=C1)SC2=CC=CC=C2[N+](=O)[O-]
231.269
231.27
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'