missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrophenyl Phenyl Sulfide 98.0+%, TCI America™
Supplier: TCI America N074425G
Specifications
| 2-Nitrophenyl Phenyl Sulfide | |
| 82°C | |
| C12H9NO2S | |
| 2-Nitrodiphenyl Sulfide, 1-Nitro-2-(phenylthio)benzene | |
| C1=CC=C(C=C1)SC2=CC=CC=C2[N+](=O)[O-] | |
| 231.269 | |
| 231.27 | |
| Crystalline Powder |
| 4171-83-9 | |
| Yellow | |
| 25 g | |
| ZPWNCSAEXUDWTN-UHFFFAOYSA-N | |
| 1-nitro-2-phenylsulfanylbenzene | |
| 348620 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4171-83-9 | |
| 231.269 | |
| 2-Nitrodiphenyl Sulfide, 1-Nitro-2-(phenylthio)benzene | |
| 1-nitro-2-phenylsulfanylbenzene |
| C12H9NO2S | |
| ZPWNCSAEXUDWTN-UHFFFAOYSA-N | |
| 348620 | |
| C1=CC=C(C=C1)SC2=CC=CC=C2[N+](=O)[O-] |
Safety and Handling
TSCA : No