missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl n-Octyl Ether 98.0+%, TCI America™

Catalog No. p-7135872
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03801G 1 g
S038010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03801G Supplier TCI America Supplier No. S03801G

Chemical Identifiers

CAS 37682-29-4
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.326
MDL Number MFCD00014693
InChI Key CXVOIIMJZFREMM-UHFFFAOYSA-N
Synonym 2-nitrophenyl octyl ether, 1-nitro-2-octyloxy benzene, octyl o-nitrophenyl ether, 2-nitrophenyl n-octyl ether, 2-octyloxy nitrobenzene, benzene, 1-nitro-2-octyloxy, 1-2-nitrophenoxy octane, 1-nitro-2-octyloxybenzene, 2-n-octyloxy nitrobenzene, npoe
PubChem CID 169952
IUPAC Name 1-nitro-2-octoxybenzene
SMILES CCCCCCCCOC1=CC=CC=C1[N+](=O)[O-]

Specifications

Boiling Point 198°C
Quantity 1 g
Formula Weight 251.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitrophenyl n-Octyl Ether [Matrix for FABMS and liquid SIMS]