missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenol 98.0+%, TCI America™

Supplier:  TCI America N021925G

 View more versions of this product

Catalog No. N021925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Nitrophenol
45°C
216°C
MFCD00011688
1663
IQUPABOKLQSFBK-UHFFFAOYSA-N
2-nitrophenol
6947
139.11
Crystalline Powder
88-75-5
Yellow
C6H5NO3
25 g
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
OC1=CC=CC=C1[N+]([O-])=O
139.11
CHEBI:16260
≥98.0% (GC,T)

Chemical Identifiers

88-75-5
139.11
IQUPABOKLQSFBK-UHFFFAOYSA-N
6947
2-nitrophenol
C6H5NO3
MFCD00011688
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
CHEBI:16260
OC1=CC=CC=C1[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0777

RTECSNumber : SM2100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions