missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrofluorene 99.0+%, TCI America™

Catalog No. p-7133675
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N020125G 25 g
N0201100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N020125G Supplier TCI America Supplier No. N020125G
Only null left

Chemical Identifiers

CAS 607-57-8
Molecular Formula C13H9NO2
Molecular Weight (g/mol) 211.22
MDL Number MFCD00001117
InChI Key XFOHWECQTFIEIX-UHFFFAOYSA-N
Synonym 2-nitrofluorene, nitrofluorene, 9h-fluorene, 2-nitro, fluorene, 2-nitro, unii-191ll4u4gz, ccris 1189, dsstox_cid_971, dsstox_rid_75898, dsstox_gsid_20971
PubChem CID 11831
ChEBI CHEBI:1224
IUPAC Name 2-nitro-9H-fluorene
SMILES [O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1

Specifications

Melting Point 155°C
Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 211.22
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrofluorene
RTECSNumber LL8225000
TSCA Yes