accessibility menu, dialog, popup

UserName

CAS RN 607-57-8

IUPAC Name: 2-nitro-9H-fluorene
Synonyms: nitrofluorene
Molecular Weight (g/mol): 211.22
Molecular Formula: C13H9NO2
InChi Key: XFOHWECQTFIEIX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)