Learn More
2-Nitrobenzyl bromide, 98+%
CAS: 3958-60-9 | C7H6BrNO2 | 216.034 g/mol
Supplier: Thermo Scientific Chemicals A1312722
Description
2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.
Solubility
Insoluble in water.
Notes
Moisture sensitive. Incompatible with alcohols, bases, amines and oxidizing agents.
Specifications
| 2-Nitrobenzyl bromide | |
| 45°C to 48°C | |
| C7H6BrNO2 | |
| 100 g | |
| 638991 | |
| Insoluble in water. | |
| C1=CC=C(C(=C1)CBr)[N+](=O)[O-] | |
| 216.034 | |
| 216.04 |
| 3958-60-9 | |
| >110°C (230°F) | |
| MFCD00007184 | |
| UN3261 | |
| 2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide | |
| HXBMIQJOSHZCFX-UHFFFAOYSA-N | |
| 1-(bromomethyl)-2-nitrobenzene | |
| 77569 | |
| ≥98% |
Chemical Identifiers
| 3958-60-9 | |
| 216.034 | |
| HXBMIQJOSHZCFX-UHFFFAOYSA-N | |
| 77569 | |
| C1=CC=C(C(=C1)CBr)[N+](=O)[O-] |
| C7H6BrNO2 | |
| MFCD00007184 | |
| 2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide | |
| 1-(bromomethyl)-2-nitrobenzene |
Safety and Handling
GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 223-558-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only