missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl bromide, 98+%

Catalog No. p-7020981
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1312706 5 g
AAA1312714 25 g
AAA1312722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1312706 Supplier Thermo Scientific Chemicals Supplier No. A1312706
Only null left

CAS: 3958-60-9 | C7H6BrNO2 | 216.034 g/mol

2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with alcohols, bases, amines and oxidizing agents.

Chemical Identifiers

CAS 3958-60-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007184
InChI Key HXBMIQJOSHZCFX-UHFFFAOYSA-N
Synonym 2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide
PubChem CID 77569
IUPAC Name 1-(bromomethyl)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)CBr)[N+](=O)[O-]

Specifications

Melting Point 45°C to 48°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 638991
Solubility Information Insoluble in water.
Formula Weight 216.04
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzyl bromide
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 223-558-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only