missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl bromide, 98+%

CAS: 3958-60-9 | C7H6BrNO2 | 216.034 g/mol

Supplier:  Thermo Scientific Chemicals A1312714

 View more versions of this product

Catalog No. AAA1312714

Only null left
Explore available promotions

Description

Description

2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with alcohols, bases, amines and oxidizing agents.
Specifications

Specifications

2-Nitrobenzyl bromide
45°C to 48°C
C7H6BrNO2
25 g
638991
Insoluble in water.
C1=CC=C(C(=C1)CBr)[N+](=O)[O-]
216.034
216.04
3958-60-9
>110°C (230°F)
MFCD00007184
UN3261
2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide
HXBMIQJOSHZCFX-UHFFFAOYSA-N
1-(bromomethyl)-2-nitrobenzene
77569
≥98%

Chemical Identifiers

3958-60-9
216.034
HXBMIQJOSHZCFX-UHFFFAOYSA-N
77569
C1=CC=C(C(=C1)CBr)[N+](=O)[O-]
C7H6BrNO2
MFCD00007184
2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide
1-(bromomethyl)-2-nitrobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 223-558-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only