missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzonitrile, 98+%

Catalog No. AAA1094106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1094106 5 g
AAA1094114 25 g
AAA1094122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1094106 Supplier Thermo Scientific Chemicals Supplier No. A1094106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 612-24-8 | C7H4N2O2 | 148.12 g/mol

ET catalysis of Au25 in intramolecular cascade reaction of 2-nitrobenzonitrile, from which 2-amniobenzamide (a precursor of significant pharmaceuticals20,21) is produced with high yield. fluorodenitration of 2-nitrobenzonitrile with Rb18F in DMSO gave an 85% yield of the 18F-labelled benzonitrile after only 20 minutes at 150°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ET catalysis of Au25 in intramolecular cascade reaction of 2-nitrobenzonitrile, from which 2-amniobenzamide (a precursor of significant pharmaceuticals20,21) is produced with high yield. fluorodenitration of 2-nitrobenzonitrile with Rb18F in DMSO gave an 85% yield of the 18F-labelled benzonitrile after only 20 minutes at 150°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 612-24-8
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD00007044
InChI Key SWBDKCMOLSUXRH-UHFFFAOYSA-N
Synonym o-nitrobenzonitrile, o-cyanonitrobenzene, benzonitrile, o-nitro, benzonitrile, 2-nitro, 2-cyanonitrobenzene, nitrobenzonitrile, unii-drc6u29fci, ccris 2326, 1-nitro-2-cyanobenzene, 2-nitrobenzenecarbonitrile
PubChem CID 11922
IUPAC Name 2-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=CC=C1C#N

Specifications

Melting Point 107°C to 111°C
Boiling Point 165°C (16 mmHg)
Odor Almond-like
Quantity 5 g
UN Number UN3439
Beilstein 638987
Solubility Information Insoluble in water.
Formula Weight 148.12
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzonitrile
Hazard Category H300-H311+H331
Hazard Statement GHS H Statement
H300-H311
Fatal if swallowed.
Toxic in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 210-301-0
RTECSNumber DI4903000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only