accessibility menu, dialog, popup

UserName

CAS RN 612-24-8

IUPAC Name: 2-nitrobenzonitrile
Synonyms: 1-nitro-2-cyanobenzene
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: SWBDKCMOLSUXRH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1C#N
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)