missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzhydrazide, 98%

CAS: 606-26-8 | C7H7N3O3 | 181.151 g/mol

Supplier:  Thermo Scientific Chemicals B2133006

 View more versions of this product

Catalog No. AAB2133006

Only null left
Explore available promotions

Description

Description

The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

Solubility
Soluble in methanol(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Nitrobenzhydrazide
119°C to 123°C
MFCD00007598
394754
Soluble in methanol(almost transparency).
C1=CC=C(C(=C1)C(=O)NN)[N+](=O)[O-]
181.151
181.15
606-26-8
C7H7N3O3
5 g
2-nitrobenzhydrazide, o-nitrobenzohydrazide, o-nitrobenzhydrazide, 2-nitro-benzoic acid hydrazide, 2-nitrobenzoic hydrazide, benzoic acid, 2-nitro-, hydrazide, benzoylhydrazine, o-nitro, 2-nitrobenzenecarbohydrazide, nitrobenzhydrazid, 2-nitrobenzoylhydrazine
LYGGDXLOJMNFBV-UHFFFAOYSA-N
2-nitrobenzohydrazide
3781218
98%

Chemical Identifiers

606-26-8
181.151
LYGGDXLOJMNFBV-UHFFFAOYSA-N
3781218
C1=CC=C(C(=C1)C(=O)NN)[N+](=O)[O-]
C7H7N3O3
MFCD00007598
2-nitrobenzhydrazide, o-nitrobenzohydrazide, o-nitrobenzhydrazide, 2-nitro-benzoic acid hydrazide, 2-nitrobenzoic hydrazide, benzoic acid, 2-nitro-, hydrazide, benzoylhydrazine, o-nitro, 2-nitrobenzenecarbohydrazide, nitrobenzhydrazid, 2-nitrobenzoylhydrazine
2-nitrobenzohydrazide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 210-110-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only