missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzhydrazide, 98%

Catalog No. p-7025695
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2133006 5 g
AAB2133014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2133006 Supplier Thermo Scientific Chemicals Supplier No. B2133006
Only null left

CAS: 606-26-8 | C7H7N3O3 | 181.151 g/mol

The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

Solubility
Soluble in methanol(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 606-26-8
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00007598
InChI Key LYGGDXLOJMNFBV-UHFFFAOYSA-N
Synonym 2-nitrobenzhydrazide, o-nitrobenzohydrazide, o-nitrobenzhydrazide, 2-nitro-benzoic acid hydrazide, 2-nitrobenzoic hydrazide, benzoic acid, 2-nitro-, hydrazide, benzoylhydrazine, o-nitro, 2-nitrobenzenecarbohydrazide, nitrobenzhydrazid, 2-nitrobenzoylhydrazine
PubChem CID 3781218
IUPAC Name 2-nitrobenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)[N+](=O)[O-]

Specifications

Melting Point 119°C to 123°C
Quantity 5 g
Beilstein 394754
Solubility Information Soluble in methanol(almost transparency).
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzhydrazide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-110-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only