accessibility menu, dialog, popup

UserName

CAS RN 606-26-8

IUPAC Name: 2-nitrobenzohydrazide
Synonyms: benzoylhydrazine, O-nitro
Molecular Weight (g/mol): 181.15
Molecular Formula: C7H7N3O3
InChi Key: LYGGDXLOJMNFBV-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results