Learn More
2-Nitro-m-xylene, 99%
CAS: 81-20-9 | C8H9NO2 | 151.165 g/mol
$81.73 - $263.03
Chemical Identifiers
| CAS | 81-20-9 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD00007181 |
| InChI Key | HDFQKJQEWGVKCQ-UHFFFAOYSA-N |
| Synonym | 2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134 |
| PubChem CID | 6672 |
| IUPAC Name | 1,3-dimethyl-2-nitrobenzene |
| SMILES | CC1=C(C(=CC=C1)C)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0554622
|
Thermo Scientific Chemicals
L0554622 |
100 g |
Each for $81.73
|
|
|||||
|
AAL0554636
|
Thermo Scientific Chemicals
L0554636 |
500 g |
Each for $263.03
|
|
|||||
Description
2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.
Solubility
Miscible with ethanol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
| 81-20-9 | |
| 151.165 | |
| HDFQKJQEWGVKCQ-UHFFFAOYSA-N | |
| 6672 | |
| CC1=C(C(=CC=C1)C)[N+](=O)[O-] |
| C8H9NO2 | |
| MFCD00007181 | |
| 2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134 | |
| 1,3-dimethyl-2-nitrobenzene |
Specifications
| 81-20-9 | |
| 1.11 | |
| 87°C (188°F) | |
| 1.522 | |
| 100 g | |
| 2046066 | |
| Miscible with ethanol. Immiscible with water. | |
| CC1=C(C(=CC=C1)C)[N+](=O)[O-] | |
| 151.165 | |
| 151.17 | |
| 2-Nitro-m-xylene |
| 14°C to 16°C | |
| 225°C to 227°C | |
| C8H9NO2 | |
| MFCD00007181 | |
| UN1665 | |
| 2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134 | |
| HDFQKJQEWGVKCQ-UHFFFAOYSA-N | |
| 1,3-dimethyl-2-nitrobenzene | |
| 6672 | |
| 99% |
Safety and Handling
GHS H Statement
H312-H332-H227
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
P210-P261-P271-P280-P302+P352-P304+P340-P312-P363-P370+P378q-P501c
H227-H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROXYLENES, LIQUID
EINECSNumber : 201-333-6
RTECSNumber : ZE4686000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only