accessibility menu, dialog, popup

UserName

CH3H3CNOO

CAS RN 81-20-9

IUPAC Name: 1,3-dimethyl-2-nitrobenzene
Synonyms: 1,3-dimethyl-2-nitrobenzene
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: HDFQKJQEWGVKCQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)