missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-m-xylene, 99%

Catalog No. AAL0554636
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0554636 500 g
AAL0554622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0554636 Supplier Thermo Scientific Chemicals Supplier No. L0554636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 81-20-9 | C8H9NO2 | 151.165 g/mol

2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

Solubility
Miscible with ethanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 81-20-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007181
InChI Key HDFQKJQEWGVKCQ-UHFFFAOYSA-N
Synonym 2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134
PubChem CID 6672
IUPAC Name 1,3-dimethyl-2-nitrobenzene
SMILES CC1=C(C(=CC=C1)C)[N+](=O)[O-]

Specifications

Melting Point 14°C to 16°C
Density 1.11
Boiling Point 225°C to 227°C
Flash Point 87°C (188°F)
Refractive Index 1.522
Quantity 500 g
UN Number UN1665
Beilstein 2046066
Solubility Information Miscible with ethanol. Immiscible with water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 2-Nitro-m-xylene
Show More Show Less
Hazard Category H227-H312+H332
Hazard Statement GHS H Statement
H312-H332-H227
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P261-P271-P280-P302+P352-P304+P340-P312-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROXYLENES, LIQUID
EINECSNumber 201-333-6
RTECSNumber ZE4686000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only