missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl Phenyl Ketone 98.0+%, TCI America™

Catalog No. B03015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03015G 5 g
B030125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B03015G Supplier TCI America Supplier No. B03015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 644-13-3
Molecular Formula C17H12O
Molecular Weight (g/mol) 232.28
MDL Number MFCD00004106
InChI Key SJNXJRVDSTZUFB-UHFFFAOYSA-N
Synonym 2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
PubChem CID 69516
IUPAC Name (naphthalen-2-yl)(phenyl)methanone
SMILES O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 82°C
Boiling Point 225°C
Quantity 5 g
Formula Weight 232.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyl Phenyl Ketone