missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthol, 99+%

Catalog No. p-171122
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180821000 100 g Glass bottle
AC180825000 500 g Glass bottle
AC180820010 1 kg Glass bottle
AC180820025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC180821000 Supplier Thermo Scientific Chemicals Supplier No. 180821000
Only null left

CAS: 135-19-3 | C10H8O | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135-19-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004067
InChI Key JWAZRIHNYRIHIV-UHFFFAOYSA-N
Synonym 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
PubChem CID 8663
ChEBI CHEBI:10432
IUPAC Name naphthalen-2-ol
SMILES C1=CC=C2C=C(C=CC2=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 120.0°C to 124.0°C
Color White
Boiling Point 285.0°C to 286.0°C
Flash Point 153°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C10H7OH
Quantity 100 g
Beilstein 06,627
Merck Index 15,6469
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol
Formula Weight 144.17
Percent Purity 99+%
Physical Form Crystals, Powder or Granules
Chemical Name or Material 2-Naphthol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 205-182-7
RTECSNumber QL2975000
TSCA TSCA

RUO – Research Use Only