CAS RN 135-19-3
2-Naphthol, 98+%
CAS: 135-19-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.173 MDL Number: MFCD00004067 InChI Key: JWAZRIHNYRIHIV-UHFFFAOYSA-N Synonym: 2-naphthol,2-naphthalenol,beta-naphthol,2-hydroxynaphthalene,betanaphthol,isonaphthol,developer bn,naphthol b,azogen developer a,beta-napthol PubChem CID: 8663 ChEBI: CHEBI:10432 IUPAC Name: naphthalen-2-ol SMILES: C1=CC=C2C=C(C=CC2=C1)O
2-Naphthol, 99+%
CAS: 135-19-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.17 MDL Number: MFCD00004067 InChI Key: JWAZRIHNYRIHIV-UHFFFAOYSA-N Synonym: 2-naphthol,2-naphthalenol,beta-naphthol,2-hydroxynaphthalene,betanaphthol,isonaphthol,developer bn,naphthol b,azogen developer a,beta-napthol PubChem CID: 8663 ChEBI: CHEBI:10432 IUPAC Name: naphthalen-2-ol SMILES: C1=CC=C2C=C(C=CC2=C1)O
2-Naphthol, 98%
CAS: 135-19-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.17 MDL Number: MFCD00004067 InChI Key: JWAZRIHNYRIHIV-UHFFFAOYSA-N Synonym: 2-naphthol,2-naphthalenol,beta-naphthol,2-hydroxynaphthalene,betanaphthol,isonaphthol,developer bn,naphthol b,azogen developer a,beta-napthol PubChem CID: 8663 ChEBI: CHEBI:10432 IUPAC Name: naphthalen-2-ol SMILES: C1=CC=C2C=C(C=CC2=C1)O
2-Naphthol 99.0+%, TCI America™
CAS: 135-19-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.173 MDL Number: MFCD00004067 InChI Key: JWAZRIHNYRIHIV-UHFFFAOYSA-N Synonym: 2-naphthol,2-naphthalenol,beta-naphthol,2-hydroxynaphthalene,betanaphthol,isonaphthol,developer bn,naphthol b,azogen developer a,beta-napthol PubChem CID: 8663 ChEBI: CHEBI:10432 IUPAC Name: naphthalen-2-ol SMILES: C1=CC=C2C=C(C=CC2=C1)O