Learn More
2-Naphthol, 98+%
CAS: 135-19-3 | C10H8O | 144.173 g/mol
Supplier: Thermo Scientific Chemicals A145640I
Description
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
Solubility
Partly miscible in water (0.76 g/L).
Notes
It is sensitive to light. Incompatible with oxidizing agents.
Specifications
| 2-Naphthol | |
| 120°C to 124°C | |
| 5.77 | |
| 153°C (307°F) | |
| C10H8O | |
| 5000 g | |
| 742134 | |
| 14,6384 | |
| Partly miscible in water (0.76g/L). | |
| C1=CC=C2C=C(C=CC2=C1)O | |
| 144.173 | |
| CHEBI:10432 | |
| ≥98% |
| 135-19-3 | |
| 1.28 | |
| 285°C to 286°C | |
| Phenol-like | |
| MFCD00004067 | |
| UN3077 | |
| Light sensitive | |
| 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol | |
| JWAZRIHNYRIHIV-UHFFFAOYSA-N | |
| naphthalen-2-ol | |
| 8663 | |
| 144.17 |
Chemical Identifiers
| 135-19-3 | |
| 144.173 | |
| JWAZRIHNYRIHIV-UHFFFAOYSA-N | |
| 8663 | |
| naphthalen-2-ol |
| C10H8O | |
| MFCD00004067 | |
| 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol | |
| CHEBI:10432 | |
| C1=CC=C2C=C(C=CC2=C1)O |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302+H332-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 205-182-7
RTECSNumber : QL2975000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only