missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthol, 98+%
Description
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
Solubility
Partly miscible in water (0.76 g/L).
Notes
It is sensitive to light. Incompatible with oxidizing agents.
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
Solubility
Partly miscible in water (0.76 g/L).
Notes
It is sensitive to light. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 120°C to 124°C |
| Density | 1.28 |
| pH | 5.77 |
| Boiling Point | 285°C to 286°C |
| Flash Point | 153°C (307°F) |
| Odor | Phenol-like |
| Quantity | 1000 g |
| UN Number | UN3077 |
| Beilstein | 742134 |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H373 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 205-182-7 |
| RTECSNumber | QL2975000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only