missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenethiol 98.0+%, TCI America™

Catalog No. p-7133576
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N002025G 25 g
N0020250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N002025G Supplier TCI America Supplier No. N002025G
Only null left

Chemical Identifiers

CAS 91-60-1
Molecular Formula C10H8S
Molecular Weight (g/mol) 160.234
MDL Number MFCD00004086
InChI Key RFCQDOVPMUSZMN-UHFFFAOYSA-N
Synonym 2-naphthalenethiol, 2-mercaptonaphthalene, 2-thionaphthol, thionaphthol, 2-naphthyl mercaptan, vulcamel tbn, 2-naphthyl thiol, renacit 1, rpa no. 2, rpa 2
PubChem CID 7058
IUPAC Name naphthalene-2-thiol
SMILES C1=CC=C2C=C(C=CC2=C1)S

Specifications

Melting Point 81°C
Color White-Yellow
Boiling Point 211°C
Quantity 25 g
Formula Weight 160.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthalenethiol
EINECSNumber (4)-0980
RTECSNumber QK3930000
TSCA Yes