accessibility menu, dialog, popup

UserName

CAS RN 91-60-1

IUPAC Name: naphthalene-2-thiol
Synonyms: 2-naphthalenethiol
Molecular Weight (g/mol): 160.23
Molecular Formula: C10H8S
InChi Key: RFCQDOVPMUSZMN-UHFFFAOYSA-N
SMILES: SC1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)