missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenesulfonic Acid Hydrate 98.0+%, TCI America™

Catalog No. p-7118392
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
B025825G 25 g
B0258500G 500 g
2 options

Catalog No. B025825G

Supplier: TCI America B025825G

Only null left

Chemical Identifiers

CAS 76530-12-6
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00149271
InChI Key KVBGVZZKJNLNJU-UHFFFAOYSA-N
Synonym naphthalene-2-sulfonic acid hydrate, 2-naphthalenesulfonic acid hydrate, 2-naphthalenesulfonic acid monohydrate, 2-naphthalenesulfonicacid, hydrate 1:1, kyselina2-naftalensulfonova, acmc-20aj4s, ksc352o8d, c10h8so3.h2o, 2-naphthalene-sulfonic acid monohydrate, naphthalene-2-sulphonic acid monohydrate
PubChem CID 20499058
IUPAC Name naphthalene-2-sulfonic acid
SMILES OS(=O)(=O)C1=CC=C2C=CC=CC2=C1

Specifications

Quantity 25 g
UN Number 2585
Formula Weight 208.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthalenesulfonic Acid Hydrate
EINECSNumber (4)-0465
RTECSNumber QK1225000
TSCA Yes