accessibility menu, dialog, popup

UserName

CAS RN 76530-12-6

IUPAC Name: naphthalene-2-sulfonic acid
Synonyms: 2-naphthalenesulfonic acid|kyselina 2-naftalensulfonova|propoxyphene napsylate anhydrous
Molecular Weight (g/mol): 208.23
Molecular Formula: C10H8O3S
InChi Key: KVBGVZZKJNLNJU-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)