missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)naphthalene, 98%

Catalog No. p-7026393
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2236803 1 g
AAB2236806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2236803 Supplier Thermo Scientific Chemicals Supplier No. B2236803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7433-79-6 | C11H10S | 174.261 g/mol

2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7433-79-6
Molecular Formula C11H10S
Molecular Weight (g/mol) 174.261
MDL Number MFCD00051615
InChI Key SSKUUDUKJMIOKQ-UHFFFAOYSA-N
Synonym 2-methylthio naphthalene, 2-methylmercaptonaphthalene, 2-methylsulfanyl naphthalene, 2-methylthionaphthalene, methyl2-naphthylsulfide, methyl 2-naphthyl sulfide, naphth-2-yl methyl thioether, naphthalene,2-methylthio, methyl naphthalen-2-yl sulfane
PubChem CID 139022
IUPAC Name 2-methylsulfanylnaphthalene
SMILES CSC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 62°C to 63°C
Odor Stench
Quantity 1 g
Beilstein 1859409
Solubility Information Slightly soluble in water
Formula Weight 174.27
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)naphthalene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only