missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzoic acid, 98+%

Catalog No. p-7080730
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1037906 5 g
AAA1037914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1037906 Supplier Thermo Scientific Chemicals Supplier No. A1037906
Only null left

CAS: 3724-10-5 | C8H8O2S | 168.21 g/mol

2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Soluble in Methanol.

Notes
Incompatible with strong oxidizing agents, acids, bases, amines. Store in a cool, dry place. Store in a tightly closed container.

Chemical Identifiers

CAS 3724-10-5
MDL Number MFCD00029934
ChEBI CHEBI:4657

Specifications

Melting Point 166°C to 168°C
Molecular Formula C21H29N3O
Quantity 5 g
Synonym disopyramide, dicorantil, rythmodan, disopiramida, ritmodan, disopyramidum, searle 703, isorythm, lispine, disopyramidum inn-latin
Solubility Information Soluble in Methanol.
InChI Key UVTNFZQICZKOEM-UHFFFAOYNA-N
SMILES CC(C)N(CCC(C1=CC=CC=C1)(C2=CC=CC=N2)C(=O)N)C(C)C
IUPAC Name 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanamide
Molecular Weight (g/mol) 339.48
PubChem CID 3114
Formula Weight 168.21
Percent Purity ≥98%
Chemical Name or Material 2- (Methylthio)benzoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only