Learn More
2-Methylthiazolidine, 98%
CAS: 24050-16-6 | C4H9NS | 103.183 g/mol
$386.60 - $386.60
Chemical Identifiers
| CAS | 24050-16-6 |
|---|---|
| Molecular Formula | C4H9NS |
| Molecular Weight (g/mol) | 103.183 |
| MDL Number | MFCD00053110 |
| InChI Key | DQMLFUMEBNHPPB-UHFFFAOYSA-N |
| PubChem CID | 32196 |
| IUPAC Name | 2-methyl-1,3-thiazolidine |
| SMILES | CC1NCCS1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2253714
|
Thermo Scientific Chemicals
B2253714 |
25 g |
Each for $386.60
|
|
|||||
Description
2-Methylthiazolidine is used as sources of low molecular-weight thiol compounds both in EATC and mouse liver homogenate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylthiazolidine is used as sources of low molecular-weight thiol compounds both in EATC and mouse liver homogenate.
Solubility
Slightly soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 24050-16-6 | |
| 103.183 | |
| DQMLFUMEBNHPPB-UHFFFAOYSA-N | |
| 2-methyl-1,3-thiazolidine |
| C4H9NS | |
| MFCD00053110 | |
| 32196 | |
| CC1NCCS1 |
Specifications
| 24050-16-6 | |
| 35°C (95°F) | |
| 1.526 | |
| 25 g | |
| 2-methylthiazolidine, thiazolidine, 2-methyl, methyl-2 thiazolidine, methyl-2 thiazolidine french, 2-methyltetrahydrothiazole, acmc-20anpm, thiazolidine,2-methyl, 4-27-00-00661 beilstein handbook reference, +/--2-methylthiazolidine, # | |
| DQMLFUMEBNHPPB-UHFFFAOYSA-N | |
| 2-methyl-1,3-thiazolidine | |
| 32196 | |
| 99% |
| 76°C to 78°C (36 mmHg) | |
| C4H9NS | |
| MFCD00053110 | |
| UN1993 | |
| Slightly soluble in water | |
| CC1NCCS1 | |
| 103.183 | |
| 103.18 | |
| 2-Methylthiazolidine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECSNumber : XJ6061000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only