accessibility menu, dialog, popup

UserName

CAS RN 24050-16-6

IUPAC Name: 2-methyl-1,3-thiazolidine
Synonyms: 2-methyl-1,3-thiazolidine
Molecular Weight (g/mol): 103.18
Molecular Formula: C4H9NS
InChi Key: DQMLFUMEBNHPPB-UHFFFAOYNA-N
SMILES: CC1NCCS1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results